Assay Details
| Gene Symbol: nme6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131597.2 | NP_571672.2 | 4 - 5 | 775 | 102 | |
| XM_021468514.1 | XP_021324189.1 | 4 - 5 | 360 | 102 | ||
| GenBank mRNA | AF241153.1 | - | 4 - 5 | 777 | 102 | |
| EH550824.1 | - | 4 - 5 | 762 | 102 | ||
| EH561410.1 | - | 1 - 2 | 369 | 102 | ||
More Information
Set Membership: |
|

nme6
Genomic Map