Assay Details
| Gene Symbol: fabp7a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131605.2 | NP_571680.1 | 3 - 4 | 404 | 79 | |
| GenBank mRNA | AB055675.1 | - | 2 - 3 | 281 | 79 | |
| AF237712.2 | - | 3 - 4 | 404 | 79 | ||
| BC055621.1 | - | 3 - 4 | 407 | 79 | ||
| BC164937.1 | - | 3 - 4 | 377 | 79 | ||
More Information
Set Membership: |
|

fabp7a
Genomic Map