Assay Details
Gene Symbol: sst2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131727.2 | NP_571802.1 | 1 - 2 | 171 | 83 | |
GenBank mRNA | AJ238017.1 | - | 1 - 2 | 170 | 83 | |
BC162704.1 | - | 1 - 2 | 159 | 83 | ||
BC162710.1 | - | 1 - 2 | 159 | 83 | ||
EH490162.1 | - | 1 - 2 | 203 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
