Assay Details
Gene Symbol: nots | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131804.1 | NP_571879.1 | 251 | 104 | ||
GenBank mRNA | AJ278269.1 | - | 251 | 104 | ||
BC086835.1 | - | 416 | 104 | |||
BC165335.1 | - | 366 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
