Assay Details
Gene Symbol: dmbx1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178163.2 | NP_835457.2 | 2 - 3 | 237 | 85 | |
GenBank mRNA | AF398526.1 | - | 2 - 3 | 237 | 85 | |
BC134894.1 | - | 2 - 3 | 315 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
