Assay Details
| Gene Symbol: ldb3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199858.3 | NP_956152.1 | 4 - 5 | 486 | 91 | |
| XM_005156272.4 | XP_005156329.1 | 5 - 6 | 540 | 91 | ||
| XM_005156273.4 | XP_005156330.1 | 5 - 6 | 586 | 91 | ||
| GenBank mRNA | BC047189.1 | - | 5 - 6 | 521 | 91 | |
| BC065846.1 | - | 4 - 5 | 491 | 91 | ||
More Information
Set Membership: |
|

ldb3b
Genomic Map