Assay Details
| Gene Symbol: ldb3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199858.3 | NP_956152.1 | 6 - 7 | 719 | 61 | |
| XM_005156272.4 | XP_005156329.1 | 7 - 8 | 773 | 61 | ||
| XM_005156273.4 | XP_005156330.1 | 7 - 8 | 819 | 61 | ||
| GenBank mRNA | BC047189.1 | - | 7 - 8 | 754 | 61 | |
| BC065846.1 | - | 6 - 7 | 724 | 61 | ||
More Information
Set Membership: |
|

ldb3b
Genomic Map