Assay Details
| Gene Symbol: hacd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199861.1 | NP_956155.1 | 6 - 7 | 706 | 72 | |
| GenBank mRNA | BC044396.1 | - | 6 - 7 | 706 | 72 | |
| BC165084.1 | - | 6 - 7 | 668 | 72 | ||
More Information
Set Membership: |
|

hacd2
Genomic Map