Assay Details
| Gene Symbol: ormdl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200093.1 | NP_956387.1 | 2 - 3 | 252 | 89 | |
| GenBank mRNA | BC059514.1 | - | 2 - 3 | 252 | 89 | |
| BC065474.1 | - | 2 - 3 | 225 | 89 | ||
| BC068376.1 | - | 2 - 3 | 247 | 89 | ||
More Information
Set Membership: |
|

ormdl1
Genomic Map