Assay Details
| Gene Symbol: hnrnpa1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200104.1 | NP_956398.1 | 4 - 5 | 609 | 87 | |
| GenBank mRNA | BC044442.1 | - | 4 - 5 | 609 | 87 | |
More Information
Set Membership: |
|

hnrnpa1b
Genomic Map