Assay Details
| Gene Symbol: rgs14a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201304.1 | NP_958461.1 | 7 - 8 | 903 | 61 | |
| XM_009307644.3 | XP_009305919.2 | 7 - 8 | 869 | 61 | ||
| GenBank mRNA | BC055659.1 | - | 7 - 8 | 903 | 61 | |
More Information
Set Membership: |
|

rgs14a
Genomic Map