Assay Details
| Gene Symbol: rplp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212743.1 | NP_997908.1 | 2 - 3 | 186 | 97 | |
| XM_021476788.1 | XP_021332463.1 | 3 - 4 | 291 | 97 | ||
| GenBank mRNA | BC059681.1 | - | 2 - 3 | 186 | 97 | |
More Information
Set Membership: |
|

rplp2
Genomic Map