Assay Details
| Gene Symbol: mpx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001351837.1 | NP_001338766.1 | 9 - 10 | 1384 | 121 | |
| NM_212779.2 | NP_997944.2 | 9 - 10 | 1384 | 121 | ||
| XM_021479113.1 | XP_021334788.1 | 9 - 10 | 1447 | 121 | ||
| GenBank mRNA | AF349034.1 | - | 9 - 10 | 121 | ||
| AF378824.1 | - | 6 - 7 | 1061 | 121 | ||
More Information
Set Membership: |
|

mpx
Genomic Map