Assay Details
| Gene Symbol: dusp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213067.1 | NP_998232.1 | 3 - 4 | 920 | 64 | |
| GenBank mRNA | BC045494.1 | - | 3 - 4 | 920 | 64 | |
| BC071309.1 | - | 3 - 4 | 925 | 64 | ||
More Information
Set Membership: |
|

dusp1
Genomic Map