Assay Details
| Gene Symbol: ripk4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213078.1 | NP_998243.1 | 2 - 3 | 609 | 71 | |
| GenBank mRNA | AF487541.1 | - | 2 - 3 | 472 | 71 | |
| BC045432.1 | - | 2 - 3 | 609 | 71 | ||
| BC165670.1 | - | 2 - 3 | 506 | 71 | ||
More Information
Set Membership: |
|

ripk4
Genomic Map