Assay Details
| Gene Symbol: nipa2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270536.1 | NP_001257465.1 | 4 - 5 | 512 | 61 | |
| NM_213125.2 | NP_998290.1 | 4 - 5 | 451 | 61 | ||
| XM_005165975.4 | XP_005166032.1 | 4 - 5 | 927 | 61 | ||
| XM_005165976.1 | XP_005166033.1 | 4 - 5 | 649 | 61 | ||
| GenBank mRNA | BC062386.1 | - | 4 - 5 | 420 | 61 | |
| DT074263.1 | - | 4 - 5 | 397 | 61 | ||
| EH457110.1 | - | 4 - 5 | 535 | 61 | ||
More Information
Set Membership: |
|

nipa2
Genomic Map