Assay Details
Gene Symbol: med21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213423.1 | NP_998588.1 | 1 - 2 | 89 | 60 | |
GenBank mRNA | BC049042.1 | - | 1 - 2 | 89 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
