Assay Details
| Gene Symbol: igbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213522.1 | NP_998687.1 | 2 - 3 | 554 | 86 | |
| XM_005165297.4 | XP_005165354.1 | 2 - 3 | 651 | 86 | ||
| GenBank mRNA | BC045313.1 | - | 2 - 3 | 554 | 86 | |
| BC165328.1 | - | 2 - 3 | 571 | 86 | ||
More Information
Set Membership: |
|

igbp1
Genomic Map