Assay Details
| Gene Symbol: cdkn1ca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002040.1 | NP_001002040.1 | 1 - 2 | 577 | 71 | |
| GenBank mRNA | BC071317.1 | - | 1 - 2 | 577 | 71 | |
| BC153451.1 | - | 3 - 4 | 975 | 71 | ||
More Information
Set Membership: |
|

cdkn1ca
Genomic Map