Assay Details
| Gene Symbol: sra1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002047.1 | NP_001002047.1 | 3 - 4 | 437 | 73 | |
| XM_005173897.3 | XP_005173954.1 | 3 - 4 | 440 | 73 | ||
| GenBank mRNA | BC071314.1 | - | 3 - 4 | 437 | 73 | |
More Information
Set Membership: |
|

sra1
Genomic Map