Assay Details
Gene Symbol: rcn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002158.1 | NP_001002158.1 | 4 - 5 | 635 | 82 | |
XM_005164012.4 | XP_005164069.1 | 4 - 5 | 703 | 82 | ||
GenBank mRNA | BC071338.1 | - | 4 - 5 | 635 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
