Assay Details
Gene Symbol: ebp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002328.1 | NP_001002328.1 | 2 - 3 | 418 | 65 | |
GenBank mRNA | BC076453.1 | - | 2 - 3 | 418 | 65 | |
BC163987.1 | - | 1 - 2 | 346 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
