Assay Details
Gene Symbol: rpp21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003530.2 | NP_001003530.1 | 3 - 4 | 338 | 129 | |
GenBank mRNA | BC078300.1 | - | 3 - 4 | 342 | 129 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
