Assay Details
Gene Symbol: med20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003585.1 | NP_001003585.1 | 3 - 4 | 648 | 56 | |
XM_009295755.3 | XP_009294030.1 | 3 - 4 | 56 | |||
GenBank mRNA | BC078327.1 | - | 3 - 4 | 648 | 56 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
