Assay Details
Gene Symbol: cplane2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004649.1 | NP_001004649.1 | 4 - 5 | 588 | 83 | |
XM_009302545.3 | XP_009300820.1 | 3 - 4 | 278 | 83 | ||
GenBank mRNA | BC080841.1 | - | 4 - 5 | 588 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
