Assay Details
Gene Symbol: gcm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005603.1 | NP_001005603.1 | 1 - 2 | 412 | 80 | |
XM_005162569.4 | XP_005162626.1 | 2 - 3 | 418 | 80 | ||
GenBank mRNA | AB099697.1 | - | 1 - 2 | 412 | 80 | |
AB175671.1 | - | 2 - 3 | 381 | 80 | ||
AY217729.1 | - | 1 - 2 | 450 | 80 | ||
AY706754.1 | - | 1 - 2 | 104 | 80 | ||
BC162304.1 | - | 1 - 2 | 410 | 80 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005162569.4 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
