Assay Details
| Gene Symbol: peds1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006052.1 | NP_001006052.1 | 4 - 5 | 523 | 79 | |
| GenBank mRNA | BC083284.1 | - | 4 - 5 | 523 | 79 | |
More Information
Set Membership: |
|

peds1
Genomic Map