Assay Details
| Gene Symbol: nudt22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006069.1 | NP_001006069.1 | 2 - 3 | 168 | 77 | |
| XM_005173118.4 | XP_005173175.1 | 2 - 3 | 236 | 77 | ||
| XM_021481006.1 | XP_021336681.1 | 1 - 2 | 364 | 77 | ||
| GenBank mRNA | BC083253.1 | - | 2 - 3 | 168 | 77 | |
More Information
Set Membership: |
|

nudt22
Genomic Map