Assay Details
| Gene Symbol: arhgef7b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008624.1 | NP_001008624.1 | 18 - 19 | 2235 | 66 | |
| XM_017354718.2 | XP_017210207.1 | 18 - 19 | 2010 | 66 | ||
| GenBank mRNA | BC086736.1 | - | 18 - 19 | 2235 | 66 | |
| BC165668.1 | - | 18 - 19 | 2154 | 66 | ||
| DQ656108.1 | - | 18 - 19 | 2021 | 66 | ||
| DQ656109.1 | - | 18 - 19 | 2189 | 66 | ||
More Information
Set Membership: |
|

arhgef7b
Genomic Map