Assay Details
| Gene Symbol: pinx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013265.2 | NP_001013283.2 | 2 - 3 | 142 | 60 | |
| XM_021468369.1 | XP_021324044.1 | 3 - 4 | 235 | 60 | ||
| XM_021468370.1 | XP_021324045.1 | 3 - 4 | 204 | 60 | ||
| GenBank mRNA | AY576808.2 | - | 2 - 3 | 142 | 60 | |
| BC054670.1 | - | 2 - 3 | 142 | 60 | ||
| BC065317.1 | - | 1 - 2 | 118 | 60 | ||
More Information
Set Membership: |
|

pinx1
Genomic Map