Assay Details
Gene Symbol: lifra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014306.2 | NP_001014328.2 | 11 - 12 | 1681 | 89 | |
GenBank mRNA | BC091862.1 | - | 11 - 12 | 1685 | 89 | |
BC165177.1 | - | 10 - 11 | 1511 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
