Assay Details
Gene Symbol: lifra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014306.2 | NP_001014328.2 | 14 - 15 | 2120 | 153 | |
GenBank mRNA | BC091862.1 | - | 14 - 15 | 2124 | 153 | |
BC165177.1 | - | 13 - 14 | 1950 | 153 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
