Assay Details
Gene Symbol: lifra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014306.2 | NP_001014328.2 | 16 - 17 | 2355 | 90 | |
GenBank mRNA | BC091862.1 | - | 16 - 17 | 2359 | 90 | |
BC165177.1 | - | 15 - 16 | 2185 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
