Assay Details
| Gene Symbol: hibch | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014316.1 | NP_001014338.1 | 9 - 10 | 790 | 64 | |
| GenBank mRNA | BC091995.1 | - | 9 - 10 | 790 | 64 | |
| BC164778.1 | - | 9 - 10 | 775 | 64 | ||
More Information
Set Membership: |
|

hibch
Genomic Map