Assay Details
| Gene Symbol: ppa1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017833.1 | NP_001017833.1 | 10 - 11 | 901 | 81 | |
| GenBank mRNA | BC092782.1 | - | 10 - 11 | 901 | 81 | |
More Information
Set Membership: |
|

ppa1b
Genomic Map