Assay Details
Gene Symbol: rgra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017877.1 | NP_001017877.1 | 2 - 3 | 402 | 62 | |
GenBank mRNA | BC092971.1 | - | 2 - 3 | 402 | 62 | |
BC165377.1 | - | 2 - 3 | 264 | 62 | ||
KT008426.1 | - | 2 - 3 | 230 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
