Assay Details
| Gene Symbol: mmp23bb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017890.1 | NP_001017890.1 | 6 - 7 | 990 | 125 | |
| GenBank mRNA | BC092954.1 | - | 6 - 7 | 990 | 125 | |
| BC164476.1 | - | 6 - 7 | 841 | 125 | ||
More Information
Set Membership: |
|

mmp23bb
Genomic Map