Assay Details
Gene Symbol: casp6a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001020497.1 | NP_001018333.1 | 3 - 4 | 181 | 99 | |
GenBank mRNA | BC094299.1 | - | 3 - 4 | 181 | 99 | |
BC164245.1 | - | 3 - 4 | 134 | 99 | ||
MG957996.1 | - | 3 - 4 | 97 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
