Assay Details
Gene Symbol: casp6a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001020497.1 | NP_001018333.1 | 4 - 5 | 344 | 112 | |
GenBank mRNA | BC094299.1 | - | 4 - 5 | 344 | 112 | |
BC164245.1 | - | 4 - 5 | 297 | 112 | ||
MG957996.1 | - | 4 - 5 | 260 | 112 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
