Assay Details
| Gene Symbol: ndufs4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001020676.1 | NP_001018512.1 | 2 - 3 | 167 | 76 | |
| GenBank mRNA | BC095734.1 | - | 2 - 3 | 167 | 76 | |
| BC155231.1 | - | 2 - 3 | 167 | 76 | ||
More Information
Set Membership: |
|

ndufs4
Genomic Map