Assay Details
| Gene Symbol: gnpnat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024374.1 | NP_001019545.1 | 2 - 3 | 248 | 97 | |
| XM_005169825.4 | XP_005169882.1 | 3 - 4 | 401 | 97 | ||
| GenBank mRNA | BC095715.1 | - | 2 - 3 | 248 | 97 | |
More Information
Set Membership: |
|

gnpnat1
Genomic Map