Assay Details
| Gene Symbol: cdh10a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039633.1 | NP_001034722.1 | 9 - 10 | 1900 | 77 | |
| GenBank mRNA | DQ303467.1 | - | 9 - 10 | 1900 | 77 | |
More Information
Set Membership: |
|

cdh10a
Genomic Map