Assay Details
| Gene Symbol: tamalin | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039924.1 | NP_001035013.1 | 2 - 3 | 323 | 97 | |
| GenBank mRNA | BC114298.1 | - | 2 - 3 | 323 | 97 | |
More Information
Set Membership: |
|

tamalin
Genomic Map