Assay Details
| Gene Symbol: shc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044973.1 | NP_001038438.1 | 3 - 4 | 462 | 62 | |
| XM_005170462.4 | XP_005170519.2 | 3 - 4 | 1384 | 62 | ||
| XM_009296255.3 | XP_009294530.1 | 3 - 4 | 616 | 62 | ||
| XM_017353157.2 | XP_017208646.1 | 2 - 3 | 1533 | 62 | ||
| XM_017353158.2 | XP_017208647.1 | 2 - 3 | 1533 | 62 | ||
More Information
Set Membership: |
|

shc2
Genomic Map