Assay Details
| Gene Symbol: batf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045059.1 | NP_001038524.1 | 4 - 5 | 427 | 65 | |
| XM_005160860.4 | XP_005160917.1 | 2 - 3 | 559 | 65 | ||
| XM_021468479.1 | XP_021324154.1 | 2 - 3 | 219 | 65 | ||
More Information
Set Membership: |
|

batf
Genomic Map