Assay Details
Gene Symbol: nsd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082551.1 | NP_001076020.1 | 14 - 15 | 2868 | 56 | |
GenBank mRNA | BC171696.1 | - | 14 - 15 | 2862 | 56 | |
DQ358102.1 | - | 13 - 14 | 2331 | 56 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
