Assay Details
| Gene Symbol: nsd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082551.1 | NP_001076020.1 | 22 - 23 | 4152 | 91 | |
| GenBank mRNA | BC115211.1 | - | 7 - 8 | 973 | 91 | |
| BC171696.1 | - | 22 - 23 | 4146 | 91 | ||
| DQ358102.1 | - | 21 - 22 | 3615 | 91 | ||
More Information
Set Membership: |
|

nsd2
Genomic Map