Assay Details
| Gene Symbol: syt10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082842.1 | NP_001076311.1 | 4 - 5 | 1291 | 74 | |
| XM_017355049.2 | XP_017210538.2 | 4 - 5 | 1529 | 74 | ||
More Information
Set Membership: |
|

syt10
Genomic Map