Assay Details
Gene Symbol: usp12a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083556.1 | NP_001077025.1 | 6 - 7 | 756 | 104 | |
GenBank mRNA | BC115286.1 | - | 6 - 7 | 756 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
