Assay Details
Gene Symbol: map2k4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001089421.1 | NP_001082890.1 | 4 - 5 | 277 | 61 | |
XM_005172951.4 | XP_005173008.1 | 3 - 4 | 628 | 61 | ||
XM_021480155.1 | XP_021335830.1 | 4 - 5 | 665 | 61 | ||
XM_021480156.1 | XP_021335831.1 | 3 - 4 | 633 | 61 | ||
GenBank mRNA | AB438979.1 | - | 4 - 5 | 277 | 61 | |
BC162777.1 | - | 4 - 5 | 353 | 61 | ||
EF489398.1 | - | 4 - 5 | 277 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
